MilliporeSigma
  • Baseline and Kinetic Circulating Tumor Cell Counts Are Prognostic Factors in a Prospective Study of Metastatic Colorectal Cancer.
Baseline and Kinetic Circulating Tumor Cell Counts Are Prognostic Factors in a Prospective Study of Metastatic Colorectal Cancer.
Diagnostics (Basel, Switzerland) (2021-04-04)
Virgílio Souza E Silva, Emne Ali Abdallah, Angelo Borsarelli Carvalho de Brito, Alexcia Camila Braun, Milena Shizue Tariki, Celso Abdon Lopes de Mello, Vinicius Fernando Calsavara, Rachel Riechelmann, Ludmilla Thomé Domingos Chinen
ABSTRACT
The discovery of predictive biomarkers in metastatic colorectal cancer (mCRC) is essential to improve clinical outcomes. Recent data suggest a potential role of circulating tumor cells (CTCs) as prognostic indicators. We conducted a follow-on analysis from a prospective study of consecutive patients with mCRC. CTC analysis was conducted at two timepoints: baseline (CTC1; before starting chemotherapy), and two months after starting treatment (CTC2). CTC isolation/quantification were completed by ISET® (Rarecells, France). CTC expressions of drug resistance-associated proteins were evaluated. Progression-free survival (PFS) and overall survival (OS) were estimated by the Kaplan-Meier method. Seventy-five patients were enrolled from May 2012 to May 2014. A CTC1 cut-off of >1.5 CTCs/mL was associated with an inferior median OS compared to lower values. A difference of CTC2-CTC1 > 5.5 CTCs/mL was associated with a reduced median PFS. By multivariate analysis, CTC1 > 1.5 CTCs/mL was an independent prognostic factor for worse OS. Multi-drug resistance protein-1 (MRP-1) expression was associated with poor median OS. CTC baseline counts, kinetics, and MRP-1 expression were predictive of clinical outcomes. Larger studies are warranted to explore the potential clinical benefit of treating mCRC patients with targeted therapeutic regimens guided by CTC findings.
Sigma-Aldrich
Anti-ERCC1 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Monoclonal Anti-TYMS antibody produced in mouse, clone 3A1, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-ABCC1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1