MilliporeSigma
  • Transcriptional programming drives Ibrutinib-resistance evolution in mantle cell lymphoma.
Transcriptional programming drives Ibrutinib-resistance evolution in mantle cell lymphoma.
Cell reports (2021-03-18)
Xiaohong Zhao, Michelle Y Wang, Huijuan Jiang, Tint Lwin, Paul M Park, Jing Gao, Mark B Meads, Yuan Ren, Tao Li, Jiao Sun, Naima Ahmed Fahmi, Satishkumar Singh, Lalit Sehgal, Xuefeng Wang, Ariosto S Silva, Eduardo M Sotomayor, Kenneth H Shain, John L Cleveland, Michael Wang, Wei Zhang, Jun Qi, Bijal D Shah, Jianguo Tao
ABSTRACT
Ibrutinib, a bruton's tyrosine kinase (BTK) inhibitor, provokes robust clinical responses in aggressive mantle cell lymphoma (MCL), yet many patients relapse with lethal Ibrutinib-resistant (IR) disease. Here, using genomic, chemical proteomic, and drug screen profiling, we report that enhancer remodeling-mediated transcriptional activation and adaptive signaling changes drive the aggressive phenotypes of IR. Accordingly, IR MCL cells are vulnerable to inhibitors of the transcriptional machinery and especially so to inhibitors of cyclin-dependent kinase 9 (CDK9), the catalytic subunit of the positive transcription elongation factor b (P-TEFb) of RNA polymerase II (RNAPII). Further, CDK9 inhibition disables reprogrammed signaling circuits and prevents the emergence of IR in MCL. Finally, and importantly, we find that a robust and facile ex vivo image-based functional drug screening platform can predict clinical therapeutic responses of IR MCL and identify vulnerabilities that can be targeted to disable the evolution of IR.
Sigma-Aldrich
Anti-RNA polymerase II subunit B1 (phospho-CTD Ser-7) Antibody, clone 4E12, culture supernatant, clone 4E12, from rat
Sigma-Aldrich
Anti-RNA polymerase II subunit B1 (phospho-CTD Ser-5) Antibody, clone 3E8, culture supernatant, clone 3E8, from rat
Sigma-Aldrich
Anti-RNA polymerase II subunit B1 (phospho CTD Ser-2), clone 3E10 (rat monoclonal)., culture supernatant, clone 3E10, from rat