MilliporeSigma
  • Heterocyclic aromatic amine level and quality characteristics of selected Harbin red sausages in the northern Chinese market.
Heterocyclic aromatic amine level and quality characteristics of selected Harbin red sausages in the northern Chinese market.
Meat science (2020-11-08)
Xiaoyu Yin, Hongzhen Du, Ming Xu, Qian Chen, Baohua Kong
ABSTRACT
The heterocyclic aromatic amine (HAA) level and quality characteristics of selected Harbin red sausages in the northern Chinese market and the possible differences between traditional sausages and conventional sausages were evaluated in this study. Four varieties of traditional sausages and four varieties of conventional sausages were selected. Compared to conventional sausages, traditional sausages had lower moisture content and higher hardness (P < 0.05). Twelve HAAs were evaluated and eight HAAs were detected. The total HAA content was as high as 360.73 ng/g. In particular, the contents of Norharman and Harman were much higher than those of the other HAAs (P < 0.05). Additionally, the HAA contents were higher in the traditional sausages than those in the conventional sausages (P < 0.05). Principal component analysis showed that traditional and conventional sausages had a good separation based on the quality characteristics and total HAA level. The results of this study will provide useful information on the industrial production of smoked meat products.
Sigma-Aldrich
2-Amino-5-phenylpyridine, AldrichCPR