MilliporeSigma
  • Precision installation of a highly efficient suicide gene safety switch in human induced pluripotent stem cells.
Precision installation of a highly efficient suicide gene safety switch in human induced pluripotent stem cells.
Stem cells translational medicine (2020-07-15)
Zhong-Dong Shi, Jason Tchao, Ling Wu, Aaron J Carman
ABSTRACT
Human pluripotent stem cells, including induced pluripotent stem cells (iPSCs) and embryonic stem cells, hold great promise for cell-based therapies, but safety concerns that complicate consideration for routine clinical use remain. Installing a "safety switch" based on the inducible caspase-9 (iCASP9) suicide gene system should offer added control over undesirable cell replication or activity. Previous studies utilized lentiviral vectors to integrate the iCASP9 system into T cells and iPSCs. This method results in random genomic insertion of the suicide switch and inefficient killing of the cells after the switch is "turned on" with a small molecule (eg, AP1903). To improve the safety and efficiency of the iCASP9 system for use in iPSC-based therapy, we precisely installed the system into a genomic safe harbor, the AAVS1 locus in the PPP1R12C gene. We then evaluated the efficiencies of different promoters to drive iCASP9 expression in human iPSCs. We report that the commonly used EF1α promoter is silenced in iPSCs, and that the endogenous promoter of the PPP1R12C gene is not strong enough to drive high levels of iCASP9 expression. However, the CAG promoter induces strong and stable iCASP9 expression in iPSCs, and activation of this system with AP1903 leads to rapid killing and complete elimination of iPSCs and their derivatives, including MSCs and chondrocytes, in vitro. Furthermore, iPSC-derived teratomas shrank dramatically or were completely eliminated after administration of AP1903 in mice. Our data suggest significant improvements on existing iCASP9 suicide switch technologies and may serve as a guide to other groups seeking to improve the safety of stem cell-based therapies.
Sigma-Aldrich
10X PCR Buffer, Optimized for routine PCR with MgCl2 included
Sigma-Aldrich
Rimiducid, ≥90% (HPLC)
Sigma-Aldrich
Simplicon® RNA Reprogramming Kit (OKSG)