MilliporeSigma
  • Growth cone-localized microtubule organizing center establishes microtubule orientation in dendrites.
Growth cone-localized microtubule organizing center establishes microtubule orientation in dendrites.
eLife (2020-07-14)
Xing Liang, Marcela Kokes, Richard D Fetter, Maria Danielle Sallee, Adrian W Moore, Jessica L Feldman, Kang Shen
ABSTRACT
A polarized arrangement of neuronal microtubule arrays is the foundation of membrane trafficking and subcellular compartmentalization. Conserved among both invertebrates and vertebrates, axons contain exclusively 'plus-end-out' microtubules while dendrites contain a high percentage of 'minus-end-out' microtubules, the origins of which have been a mystery. Here we show that in Caenorhabditis elegans the dendritic growth cone contains a non-centrosomal microtubule organizing center (MTOC), which generates minus-end-out microtubules along outgrowing dendrites and plus-end-out microtubules in the growth cone. RAB-11-positive endosomes accumulate in this region and co-migrate with the microtubule nucleation complex γ-TuRC. The MTOC tracks the extending growth cone by kinesin-1/UNC-116-mediated endosome movements on distal plus-end-out microtubules and dynein clusters this advancing MTOC. Critically, perturbation of the function or localization of the MTOC causes reversed microtubule polarity in dendrites. These findings unveil the endosome-localized dendritic MTOC as a critical organelle for establishing axon-dendrite polarity.
Product Description
Supelco
Levamisol hydrochloride, VETRANAL®, analytical standard