MilliporeSigma
Potassium starvation induces autophagy in yeast.
The Journal of biological chemistry (2020-08-14)
Nambirajan Rangarajan, Ishani Kapoor, Shuang Li, Peter Drossopoulos, Kristen K White, Victoria J Madden, Henrik G Dohlman
ABSTRACT
Autophagy is a conserved process that recycles cellular contents to promote survival. Although nitrogen limitation is the canonical inducer of autophagy, recent studies have revealed several other nutrients important to this process. In this study, we used a quantitative, high-throughput assay to identify potassium starvation as a new and potent inducer of autophagy in the yeast Saccharomyces cerevisiae We found that potassium-dependent autophagy requires the core pathway kinases Atg1, Atg5, and Vps34, and other components of the phosphatidylinositol 3-kinase complex. Transmission EM revealed abundant autophagosome formation in response to both stimuli. RNA-Seq indicated distinct transcriptional responses: nitrogen affects transport of ions such as copper, whereas potassium targets the organization of other cellular components. Thus, nitrogen and potassium share the ability to influence molecular supply and demand but do so in different ways. Both inputs promote catabolism through bulk autophagy, but result in distinct mechanisms of cellular remodeling and synthesis.
Sigma-Aldrich
1-Naphthyl phosphate disodium salt, phosphatase substrate