MilliporeSigma
  • Loss of STI1-mediated neuronal survival and differentiation in disease-associated mutations of prion protein.
Loss of STI1-mediated neuronal survival and differentiation in disease-associated mutations of prion protein.
Journal of neurochemistry (2018-01-18)
Michele Christine Landemberger, Gabriela Pintar de Oliveira, Cleiton Fagundes Machado, Kenneth John Gollob, Vilma Regina Martins
ABSTRACT
Cellular prion protein (PrPC ) is widely expressed and displays a variety of well-described functions in the central nervous system (CNS). Mutations of the PRNP gene are known to promote genetic human spongiform encephalopathies, but the components of gain- or loss-of-function mutations to PrPC remain a matter for debate. Among the proteins described to interact with PrPC is Stress-inducible protein 1 (STI1), a co-chaperonin that is secreted from astrocytes and triggers neuroprotection and neuritogenesis through its interaction with PrPC . In this work, we evaluated the impact of different PrPC pathogenic point mutations on signaling pathways induced by the STI1-PrPC interaction. We found that some of the pathogenic mutations evaluated herein induce partial or total disruption of neuritogenesis and neuroprotection mediated by mitogen-activated protein kinase (MAPK)/extracellular signal-regulated kinases 1 and 2 (ERK1/2) and protein kinase A (PKA) signaling triggered by STI1-PrPC engagement. A pathogenic mutant PrPC that lacked both neuroprotection and neuritogenesis activities fail to promote negative dominance upon wild-type PrPC . Also, a STI1-α7-nicotinic acetylcholine receptor-dependent cellular signaling was present in a PrPC mutant that maintained both neuroprotection and neuritogenesis activities similar to what has been previously observed by wild-type PrPC . These results point to a loss-of-function mechanism underlying the pathogenicity of PrPC mutations.
Sigma-Aldrich
Anti-Prion Protein Antibody, a.a. 109-112, clone 3F4, clone 3F4, Chemicon®, from mouse