MilliporeSigma
  • Genome-Wide Chromatin Analysis of FFPE Tissues Using a Dual-Arm Robot with Clinical Potential.
Genome-Wide Chromatin Analysis of FFPE Tissues Using a Dual-Arm Robot with Clinical Potential.
Cancers (2021-05-01)
Syuzo Kaneko, Toutai Mitsuyama, Kouya Shiraishi, Noriko Ikawa, Kanto Shozu, Ai Dozen, Hidenori Machino, Ken Asada, Masaaki Komatsu, Asako Kukita, Kenbun Sone, Hiroshi Yoshida, Noriko Motoi, Shinya Hayami, Yutaka Yoneoka, Tomoyasu Kato, Takashi Kohno, Toru Natsume, Gottfried von Keudell, Vassiliki Saloura, Hiroki Yamaue, Ryuji Hamamoto
ABSTRACT
Although chromatin immunoprecipitation and next-generation sequencing (ChIP-seq) using formalin-fixed paraffin-embedded tissue (FFPE) has been reported, it remained elusive whether they retained accurate transcription factor binding. Here, we developed a method to identify the binding sites of the insulator transcription factor CTCF and the genome-wide distribution of histone modifications involved in transcriptional activation. Importantly, we provide evidence that the ChIP-seq datasets obtained from FFPE samples are similar to or even better than the data for corresponding fresh-frozen samples, indicating that FFPE samples are compatible with ChIP-seq analysis. H3K27ac ChIP-seq analyses of 69 FFPE samples using a dual-arm robot revealed that driver mutations in EGFR were distinguishable from pan-negative cases and were relatively homogeneous as a group in lung adenocarcinomas. Thus, our results demonstrate that FFPE samples are an important source for epigenomic research, enabling the study of histone modifications, nuclear chromatin structure, and clinical data.
Sigma-Aldrich
SeqPlex DNA Amplification Kit, For use with high throughput sequencing technologies, Whole Genome Amplification kit designed to facilitate Next Gen Sequencing.