MilliporeSigma
  • A Randomized Phase II Trial of Short-Course Androgen Deprivation Therapy With or Without Bevacizumab for Patients With Recurrent Prostate Cancer After Definitive Local Therapy.
A Randomized Phase II Trial of Short-Course Androgen Deprivation Therapy With or Without Bevacizumab for Patients With Recurrent Prostate Cancer After Definitive Local Therapy.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology (2016-04-06)
Rana R McKay, Amado J Zurita, Lillian Werner, Justine Y Bruce, Michael A Carducci, Mark N Stein, Elisabeth I Heath, Arif Hussain, Hai T Tran, Christopher J Sweeney, Robert W Ross, Philip W Kantoff, Susan F Slovin, Mary-Ellen Taplin
ABSTRACT
Patients with recurrent prostate cancer after local treatment make up a heterogeneous population for whom androgen deprivation therapy (ADT) is the usual treatment. The purpose of this randomized phase II trial was to investigate the efficacy and toxicity of short-course ADT with or without bevacizumab in men with hormone-sensitive prostate cancer. Eligible patients had an increasing prostate-specific antigen (PSA) of ≤ 50 ng/mL and PSA doubling time of less than 18 months. Patients had either no metastases or low burden, asymptomatic metastases (lymph nodes < 3 cm and five or fewer bone metastases). Patients were randomly assigned 2:1 to a luteinizing hormone-releasing hormone agonist, bicalutamide and bevacizumab or ADT alone, for 6 months. The primary end point was PSA relapse-free survival (RFS). Relapse was defined as a PSA of more than 0.2 ng/mL for prostatectomy patients or PSA of more than 2.0 ng/mL for primary radiation therapy patients. Sixty-six patients received ADT + bevacizumab and 36 received ADT alone. Patients receiving ADT + bevacizumab had a statistically significant improvement in RFS compared with patients treated with ADT alone (13.3 months for ADT + bevacizumab v 10.2 months for ADT alone; hazard ratio, 0.47; 95% CI, 0.29 to 0.77; log-rank P = .002). Hypertension was the most common adverse event in patients receiving ADT + bevacizumab (36%). ADT combined with bevacizumab resulted in an improved RFS for patients with hormone-sensitive prostate cancer. Long-term follow-up is needed to determine whether some patients have a durable PSA response and are able to remain off ADT for prolonged periods. Our data provide rationale for combining vascular endothelial growth factor-targeting therapy with ADT in hormone-sensitive prostate cancer.
Product Description
Millipore
MILLIPLEX® Human High Sensitivity T Cell Panel Premixed 13-plex - Immunology Multiplex Assay, Simultaneous analyze low levels of cytokine and chemokine biomarker with the High Sensitivity Bead-Based Multiplex Assays using the Luminex technology, in human serum, plasma and cell culture samples.