MilliporeSigma
  • Inflammatory biomarkers and spontaneous preterm birth among obese women.
Inflammatory biomarkers and spontaneous preterm birth among obese women.
The journal of maternal-fetal & neonatal medicine : the official journal of the European Association of Perinatal Medicine, the Federation of Asia and Oceania Perinatal Societies, the International Society of Perinatal Obstetricians (2015-12-25)
Matthew B Wallenstein, Laura L Jelliffe-Pawlowski, Wei Yang, Suzan L Carmichael, David K Stevenson, Kelli K Ryckman, Gary M Shaw
ABSTRACT
To identify associations between second-trimester serum inflammatory biomarkers and preterm birth among obese women. In this nested case-control study, we compared 65 serum inflammatory biomarkers in obese women whose pregnancies resulted in early spontaneous preterm birth (<32 weeks gestation, n = 34) to obese women whose pregnancies resulted in term birth (n = 34). These women were selected from a larger population-based California cohort. Random forest and classification and regression tree techniques were employed to identify biomarkers of importance, and adjusted odds ratios (aORs) and 95% confidence intervals (CI) were estimated using logistic regression. Random forest and classification and regression tree techniques found that soluble vascular endothelial growth factor receptor-3 (sVEGFR3), soluble interleukin-2 receptor alpha-chain (sIL-2RA) and soluble tumor necrosis factor receptor-1 (sTNFR1) were related to preterm birth. Using multivariable logistic regression to compare preterm cases and term controls, decreased serum levels of sVEGFR3 and increased serum levels of sIL-2RA and sTNFR1 were associated with increased risk of preterm birth among obese women, aOR = 3.2 (95% CI: 1.0-9.9), aOR = 2.8 (95% CI: 0.9-9.0), and aOR = 4.1 (95% CI: 1.2-14.1), respectively. In this pilot study, we identified three serum biomarkers indicative of inflammation to be associated with spontaneous preterm birth among obese women: sVEGFR3, sIL-2RA and sTNFR1.
Product Description
Millipore
MILLIPLEX® Human Soluble Cytokine Receptor Panel, HSCRMAG-32K- Immunology Multiplex Assay, Simultaneously analyze multiple cytokine and chemokine biomarkers with Bead-Based Multiplex Assays using the Luminex technology, in human serum, plasma and cell culture samples.
Millipore
MILLIPLEX® Human Adipokine Magnetic Bead Panel 1 - Endocrine Multiplex Assay, The Human Adipokine Panel 1, using the Luminex xMAP technology, enables the simultaneous analysis of five biomarkers in human serum or plasma samples.