MilliporeSigma
  • Surface Co-presentation of BMP-2 and integrin selective ligands at the nanoscale favors α5β1 integrin-mediated adhesion.
Surface Co-presentation of BMP-2 and integrin selective ligands at the nanoscale favors α5β1 integrin-mediated adhesion.
Biomaterials (2020-11-04)
Francesca Posa, Elisabeth H Baha-Schwab, Qiang Wei, Adriana Di Benedetto, Stefanie Neubauer, Florian Reichart, Horst Kessler, Joachim P Spatz, Corinne Albiges-Rizo, Giorgio Mori, Elisabetta Ada Cavalcanti-Adam
ABSTRACT
Here we present the use of surface nanopatterning of covalently immobilized BMP-2 and integrin selective ligands to determine the specificity of their interactions in regulating cell adhesion and focal adhesion assembly. Gold nanoparticle arrays carrying single BMP-2 dimers are prepared by block-copolymer micellar nanolithography and azide-functionalized integrin ligands (cyclic-RGD peptides or α5β1 integrin peptidomimetics) are immobilized on the surrounding polyethylene glycol alkyne by click chemistry. Compared to BMP-2 added to the media, surface immobilized BMP-2 (iBMP-2) favors the spatial segregation of adhesion clusters and enhances focal adhesion (FA) size in cells adhering to α5β1 integrin selective ligands. Moreover, iBMP-2 copresented with α5β1 integrin ligands induces the recruitment of αvβ3 integrins in FAs. When copresented with RGD, iBMP-2 induces the assembly of a higher number of FAs, which are not affected by α5β1 integrin blocking. Our dual-functionalized platforms offer the possibility to study the crosstalk between integrins and BMP receptors, and more in general they could be used to address the spatial regulation of growth factors and adhesion receptors crosstalk on biomimetic surfaces.
Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone hVIN-1, ascites fluid
Sigma-Aldrich
Vitronectin from human plasma, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Fibronectin bovine plasma, solution, sterile-filtered, BioReagent, suitable for cell culture