MilliporeSigma
  • Mitochondrial respiratory chain inhibition and Na+K+ATPase dysfunction are determinant factors modulating the toxicity of nickel in the brain of indian catfish Clarias batrachus L.
Mitochondrial respiratory chain inhibition and Na+K+ATPase dysfunction are determinant factors modulating the toxicity of nickel in the brain of indian catfish Clarias batrachus L.
Interdisciplinary toxicology (2019-11-26)
Arpan Kumar Maiti, Nimai Chandra Saha, Goutam Paul, Kishore Dhara
ABSTRACT
Nickel is a potential neurotoxic pollutant inflicting damage in living organisms, including fish, mainly through oxidative stress. Previous studies have demonstrated the impact of nickel toxicity on mitochondrial function, but there remain lacunae on the damage inflicted at mitochondrial respiratory level. Deficient mitochondrial function usually affects the activities of important adenosinetriphosphatases responsible for the maintenance of normal neuronal function, namely Na+K+ATPase, as explored in our study. Previous reports demonstrated the dysfunction of this enzyme upon nickel exposure but the contributing factors for the inhibition of this enzyme remained unexplored. The main purpose of this study was to elucidate the impact of nickel neurotoxicity on mitochondrial respiratory complexes and Na+K+ATPase in the piscine brain and to determine the contributing factors that had an impact on the same. Adult Clarias batrachus were exposed to nickel treated water at 10% and 20% of the 96 h LC50 value (41 mg.l-1) respectively and sampled on 20, 40 and 60 days. Exposure of fish brain to nickel led to partial inhibition of complex IV of mitochondrial respiratory chain, however, the activities of complex I, II and III remained unaltered. This partial inhibition of mitochondrial respiratory chain might have been sufficient to lower mitochondrial energy production in mitochondria that contributed to the partial dysfunction of Na+K+ATPase. Besides energy depletion other contributing factors were involved in the dysfunction of this enzyme, like loss of thiol groups for enzyme activity and lipid peroxidation-derived end products that might have induced conformational and functional changes. However, providing direct evidence for such conformational and functional changes of Na+K+ATPase was beyond the scope of the present study. In addition, immunoblotting results also showed a decrease in Na+K+ATPase protein expression highlighting the impact of nickel neurotoxicity on the expression of the enzyme itself. The implication of the inhibition of mitochondrial respiration and Na+K+ATPase dysfunction was the neuronal death as evidenced by enhanced caspase-3 and caspase-9 activities. Thus, this study established the deleterious impact of nickel neurotoxicity on mitochondrial functions in the piscine brain and identified probable contributing factors that can act concurrently in the inhibition of Na+K+ATPase. This study also provided a vital clue about the specific areas that the therapeutic agents should target to counter nickel neurotoxicity.
Sigma-Aldrich
Isolated Mitochondria Staining Kit, 1 kit sufficient for 50 reactions (in a 2 mL cuvette), 1 kit sufficient for 1000 reactions (using 96 multiwell plates)