MilliporeSigma
A stele-enriched gene regulatory network in the Arabidopsis root.
Molecular systems biology (2011-01-20)
Siobhan M Brady, Lifang Zhang, Molly Megraw, Natalia J Martinez, Eric Jiang, Charles S Yi, Weilin Liu, Anna Zeng, Mallorie Taylor-Teeples, Dahae Kim, Sebastian Ahnert, Uwe Ohler, Doreen Ware, Albertha J M Walhout, Philip N Benfey
ABSTRACT
Tightly controlled gene expression is a hallmark of multicellular development and is accomplished by transcription factors (TFs) and microRNAs (miRNAs). Although many studies have focused on identifying downstream targets of these molecules, less is known about the factors that regulate their differential expression. We used data from high spatial resolution gene expression experiments and yeast one-hybrid (Y1H) and two-hybrid (Y2H) assays to delineate a subset of interactions occurring within a gene regulatory network (GRN) that determines tissue-specific TF and miRNA expression in plants. We find that upstream TFs are expressed in more diverse cell types than their targets and that promoters that are bound by a relatively large number of TFs correspond to key developmental regulators. The regulatory consequence of many TFs for their target was experimentally determined using genetic analysis. Remarkably, molecular phenotypes were identified for 65% of the TFs, but morphological phenotypes were associated with only 16%. This indicates that the GRN is robust, and that gene expression changes may be canalized or buffered.
Sigma-Aldrich
REDExtract-N-Amp Plant PCR Kit, sufficient for 100 extractions, sufficient for 100 amplifications