MilliporeSigma
  • Natural epiestriol-16 act as potential lead molecule against prospective molecular targets of multidrug resistant Acinetobacter baumannii-Insight from in silico modelling and in vitro investigations.
Natural epiestriol-16 act as potential lead molecule against prospective molecular targets of multidrug resistant Acinetobacter baumannii-Insight from in silico modelling and in vitro investigations.
Infection, genetics and evolution : journal of molecular epidemiology and evolutionary genetics in infectious diseases (2020-04-09)
Sinosh Skariyachan, Aditi G Muddebihalkar, Vaishnavi Badrinath, Bindu Umashankar, Daniya Eram, Akshay Uttarkar, Vidya Niranjan
ABSTRACT
The current study aimed to identify putative drug targets of multidrug resistant Acinetobacter baumannii (MDRAb) and study the therapeutic potential of natural epiestriol-16 by computer aided virtual screening and in vitro studies. The clinical isolates (n = 5) showed extreme dug resistance to carbapenems and colistins (p ≤ .05). Computational screening suggested that out of 236 natural molecules selected, 06 leads were qualified for drug likeliness, pharmacokinetic features and one potential molecule namely natural epiestriol-16 (16b-Hydroxy-17a-estradiol) exhibited significant binding potential towards four prioritised drug targets in comparison with the binding of faropenem to their usual target. Natural epiestriol demonstrated profound binding to the outer membrane protein (Omp38), protein RecA (RecA), orotate phosphoribosyltransferase (PyrE) and orotidine 5'-phosphate decarboxylase (PyrF) with binding energy of -6.0, -7.3, -7.3 and -8.0 kcal/mol respectively. MD simulations suggested that 16-epiestriol-receptor complexes demonstrated stability throughout the simulation. The growth curve and time kill assays revealed that MDRAb showed resistance to faropenem and polymyxin-B and the pure epiestriol-16 showed significant inhibitory properties at a concentration of 200 μg/mL (p ≤ .5). Thus, natural epiestriol-16 can be used as potential inhibitor against the prioritised targets of MDRAb and this study provide insight for drug development against carbapenem and colistin resistant A. baumannii.
Sigma-Aldrich
16-Epiestriol, ≥93% (HPLC)