MilliporeSigma
  • Comparative genome profiling across subtypes of low-grade B-cell lymphoma identifies type-specific and common aberrations that target genes with a role in B-cell neoplasia.
Comparative genome profiling across subtypes of low-grade B-cell lymphoma identifies type-specific and common aberrations that target genes with a role in B-cell neoplasia.
Haematologica (2008-03-28)
Bibiana I Ferreira, Juan F García, Javier Suela, Manuela Mollejo, Francisca I Camacho, Angel Carro, Santiago Montes, Miguel A Piris, Juan C Cigudosa
ABSTRACT
Low-grade B-cell lymphomas are a very heterogeneous group of tumors, whose differential diagnosis is frequently compromised by the lack of specific cytogenetic or molecular features. Our objective was to search for genomic features that allow a better molecular identification of the different types of lymphoma studied. We selected a panel of 87 low-grade B-cell lymphoma tumor samples that were unambiguously diagnosed (clinically and cytogenetically) as: follicular, splenic marginal zone, nodal marginal zone, lymphoplasmacytic, mantle cell, extranodal marginal zone MALT-type lymphoma or B-cell chronic lymphocytic leukemia. All samples were subjected to the same high-resolution genomic DNA analysis (array-based comparative genomic hybridization): a whole genome platform that contained 44000 probes distributed across the genome. Genomic imbalances were recorded, compiled and analyzed. Eighty percent of analyzed cases showed genomic imbalances (deletions and gain/amplifications) but the frequency of these imbalances ranged from 100% in mantle cell lymphomas to 33% in MALT lymphomas. A total of 95 new genomic imbalances affecting all lymphoma subtypes, were defined. We evaluated the extension of the genomic instability, detecting distinct patterns of genomic instability within subtypes. Specific pathways, such as nuclear factor kB (gains of REL and BCL11A, and losses of COMMD3, BIRC1, IKK1 and NFKB2), Polycomb group proteins (gain of BMI1 and deletion of PCGF6), DNA repair checkpoint pathways (deletion of 16q24 involving CDT1), or miRNA with a role in B-cell lymphoma pathogenesis (MIRN15A, MIRN16-1), were targeted by this genomic instability. Although all subtypes of lymphomas showed gains and losses of DNA, the analysis of their genomic profiles indicated that there are specific aberrations in almost every subtype as well as frequent aberrations that are common to a large number of lymphoma types. These common aberrations target genes that are important in B-cell lymphomagenesis.