MilliporeSigma
  • Dedifferentiation-associated changes in morphology and gene expression in primary human articular chondrocytes in cell culture.
Dedifferentiation-associated changes in morphology and gene expression in primary human articular chondrocytes in cell culture.
Osteoarthritis and cartilage (2002-02-14)
M Schnabel, S Marlovits, G Eckhoff, I Fichtel, L Gotzen, V Vécsei, J Schlegel
ABSTRACT
The aim of the present study was the investigation of differential gene expression in primary human articular chondrocytes (HACs) and in cultivated cells derived from HACs. Primary human articular chondrocytes (HACs) isolated from non-arthritic human articular cartilage and monolayer cultures of HACs were investigated by immunohistochemistry, Northern analysis, RT-PCR and cDNA arrays. By immunohistochemistry we detected expression of collagen II, protein S-100, chondroitin-4-sulphate and vimentin in freshly isolated HACs. Cultivated HACs, however, showed only collagen I and vimentin expression. These data were corroborated by the results of Northern analysis using specifc cDNA probes for collagens I, II and III and chondromodulin, respectively, demonstrating collagen II and chondromodulin expression in primary HACs but not in cultivated cells. Hybridization of mRNA from primary HACs and cultivated cells to cDNA arrays revealed additional transcriptional changes associated with dedifferentiation during propagation of chondrocytes in vitro. We found a more complex hybridization pattern for primary HACs than for cultivated cells. Of the genes expressed in primary HACs the early growth response (EGR1) transcription factor showed the strongest expression whereas D-type cyclin was expressed in proliferating cells. Other factors associated with differentiated HACs were the adhesion molecules ICAM-1 and VCAM-1, VEGF, TGFbeta2, and the monocyte chemotactic protein receptor. Our data support the hypothesis that HACs dedifferentiate when grown in monolayer cultures. Moreover, the expression patterns also show that proliferation and differentiation are exclusive features of human chondrocytes.
Sigma-Aldrich
Anti-Chondroitin 4 Sulfate Antibody, clone BE-123, ascites fluid, clone BE-123, Chemicon®