MilliporeSigma
  • Acid-base characteristics of bromophenol blue-citrate buffer systems in the amorphous state.
Acid-base characteristics of bromophenol blue-citrate buffer systems in the amorphous state.
Journal of pharmaceutical sciences (2004-02-06)
Jinjiang Li, Koustuv Chatterjee, Ales Medek, Evgenyi Shalaev, George Zografi
ABSTRACT
In this study, we have examined the acid-base characteristics of various citrate buffer systems alone and in the presence of the pH indicator dye, bromophenol blue, in aqueous solution, and after lyophilization to produce amorphous material. Fourier transform Raman and solid-state nuclear magnetic resonance spectroscopy have been used to monitor the ratio of ionized to un-ionized citric acid under various conditions, as a function of initial pH in the range of 2.65-4.28. Ultraviolet-visible spectrophotometry was used to probe the extent of proton transfer of bromophenol blue in the citrate buffer systems in solution and the amorphous state. Spectroscopic studies indicated greater ionization of citric acid and bromophenol blue in solution and the solid state with increasing initial solution pH, as expected. Fourier transform Raman measurements indicated the same ratio of ionized to un-ionized citrate species in solution, frozen solution, and the amorphous state. It is shown that the ratio of species at any particular initial pH is primarily determined by the amount of sodium ion present so as to maintain electroneutrality and not necessarily to the fact that pH and pK(a) remain unchanged during freezing and freeze drying. Indeed, for bromophenol blue, the relative ultraviolet-visible intensities for ionized and un-ionized species in the amorphous sample were different from those in solution indicating that the extent of protonation of bromophenol blue was significantly lower in the solid samples. It is concluded that under certain conditions there can be significant differences in the apparent hydrogen activity of molecules in amorphous systems.
Sigma-Aldrich
Bromophenol Blue sodium salt, Dye content 90 %, ACS reagent