MilliporeSigma
  • Compulsive alcohol consumption is regulated by dorsal striatum fast-spiking interneurons.
Compulsive alcohol consumption is regulated by dorsal striatum fast-spiking interneurons.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology (2020-07-15)
Michael S Patton, Morgan Heckman, Cecelia Kim, Chaoqi Mu, Brian N Mathur, Michael S Patton, Morgan Heckman, Cecelia Kim, Chaoqi Mu, Brian N Mathur
ABSTRACT
Compulsive alcohol consumption is a core, treatment-resistant feature of alcohol use disorder. The dorsomedial and dorsolateral striatum support goal-directed and habitual action strategies, respectively. How ethanol targets dorsolateral striatum to drive compulsive consumption is poorly understood. Parvalbumin-expressing striatal fast-spiking interneurons comprise ~1% of the total neuronal striatal population, are enriched dorsolaterally and are functionally modulated by ethanol. To test whether fast-spiking interneurons are necessary for the development of compulsive ethanol consumption, we selectively ablated these neurons in adult male and female C57BL/6 J mice undergoing a voluntary chronic intermittent ethanol consumption paradigm followed by a compulsive ethanol drinking assay. Fast-spiking interneuron ablation curtailed the development of organized ethanol lick sequence behavior, reduced ethanol consumption, and abrogated compulsive consumption of ethanol with the added bitterant quinine. In contrast, fast-spiking interneuron ablation did not affect any index of water or sucrose consumption. These data causally implicate the minority striatal fast-spiking interneuron population as a key component of compulsive ethanol consumption.
Sigma-Aldrich
Ethanol Assay Kit, sufficient for 100 colorimetric or fluorometric tests