MilliporeSigma
  • Accumulation of elements by submerged (Stuckenia pectinata (L.) Börner) and emergent (Phragmites australis (Cav.) Trin. ex Steud.) macrophytes under different salinity levels.
Accumulation of elements by submerged (Stuckenia pectinata (L.) Börner) and emergent (Phragmites australis (Cav.) Trin. ex Steud.) macrophytes under different salinity levels.
Plant physiology and biochemistry : PPB (2020-06-28)
O V Anishchenko, A P Tolomeev, E A Ivanova, A V Drobotov, A A Kolmakova, I V Zuev, I V Gribovskaya
ABSTRACT
Accumulation of essential/beneficial and non-essential chemical elements by submerged (Stuckenia pectinata (L.) Börner) and emergent (Phragmites australis (Cav.) Trin. ex Steud.) rooted macrophytes in three water bodies was studied as dependent on water salinity (specific conductivity) and main environmental factors (temperature, pH of water, growing period, element contents in water and sediments). Inductively coupled plasma optical emission spectrometry (ICP - OES) was used for the element detection in the plants, sediments, and water. Multivariate statistics redundancy analysis (RDA) showed that investigated factors together explained 37.8 - 44.7% and 29.9 - 42.6% of the essential/beneficial and non-essential elements variability, respectively, whereas salinity was one of the main factors determining the accumulation of a number of elements (Mg, S, Na, Li, Sr, Sb) by the macrophytes. A decrease in the essential element contents (K, P, N, Cu, Zn) was observed in both species of the macrophytes during the growing season. Environmental factors (salinity, growing period, and pH) were more significant than the element contents in water and the sediments for the essential/beneficial element accumulation by two species of the macrophytes. The element contents in the sediments to a greater extent determined non-essential element accumulation by P. australis than by S. pectinata. The study showed that the efficiency of using P. australis and S. pectinata for remediation of aquatic ecosystems contaminated by heavy metals should not decrease with a salinity increase.
Product Description
Supelco
Scandium Standard for ICP, TraceCERT®, 10 g/L Sc in nitric acid (nominal concentration)