MilliporeSigma
  • PIKfyve, a class III PI kinase, is the target of the small molecular IL-12/IL-23 inhibitor apilimod and a player in Toll-like receptor signaling.
PIKfyve, a class III PI kinase, is the target of the small molecular IL-12/IL-23 inhibitor apilimod and a player in Toll-like receptor signaling.
Chemistry & biology (2013-07-31)
Xinming Cai, Yongyao Xu, Atwood K Cheung, Ronald C Tomlinson, Abel Alcázar-Román, Leon Murphy, Andreas Billich, Bailin Zhang, Yan Feng, Martin Klumpp, Jean-Michel Rondeau, Aleem N Fazal, Christopher J Wilson, Vic Myer, Gerard Joberty, Tewis Bouwmeester, Mark A Labow, Peter M Finan, Jeffrey A Porter, Hidde L Ploegh, Daniel Baird, Pietro De Camilli, John A Tallarico, Qian Huang
ABSTRACT
Toll-like receptor (TLR) signaling is a key component of innate immunity. Aberrant TLR activation leads to immune disorders via dysregulation of cytokine production, such as IL-12/IL-23. Herein, we identify and characterize PIKfyve, a lipid kinase, as a critical player in TLR signaling using apilimod as an affinity tool. Apilimod is a potent small molecular inhibitor of IL-12/IL-23 with an unknown target and has been evaluated in clinical trials for patients with Crohn's disease or rheumatoid arthritis. Using a chemical genetic approach, we show that it binds to PIKfyve and blocks its phosphotransferase activity, leading to selective inhibition of IL-12/IL-23p40. Pharmacological or genetic inactivation of PIKfyve is necessary and sufficient for suppression of IL-12/IL-23p40 expression. Thus, we have uncovered a phosphoinositide-mediated regulatory mechanism that controls TLR signaling.
Sigma-Aldrich
Apilimod, ≥98% (HPLC)