MilliporeSigma
  • Period of Boar Ejaculate Collection Contributes to the Yearly Intra-Male Variability of Seminal Plasma Cytokines.
Period of Boar Ejaculate Collection Contributes to the Yearly Intra-Male Variability of Seminal Plasma Cytokines.
Biology (2020-05-24)
Lorena Padilla, Xiomara Lucas, Inmaculada Parrilla, Cristina Perez-Patiño, Heriberto Rodriguez-Martinez, Jordi Roca, Isabel Barranco
ABSTRACT
The concentrations of cytokines in seminal plasma (SP) fluctuate over time in healthy males, weakening their practical usefulness as diagnostic tools. This study evaluated the relevance of intra-male variability in SP cytokines and to what extent the period of the year when ejaculate is collected contributes to such variability. Thirteen cytokines (GM-CSF, IFNγ, IL-1α, IL-1β, IL-1ra, IL-2, IL-4, IL-6, IL-8, IL-10, IL-12, IL-18, and TNFα) were measured using the Luminex xMAP® technology for 180 SP samples of ejaculate collected over a year from nine healthy and fertile boars. The SP samples were grouped into two annual periods according to decreasing or increasing daylight and ambient temperature. Intra-male variability was higher than inter-male variability for all cytokines. All SP cytokines showed concentration differences between the two periods of the year, showing the highest concentration during the increasing daylength/temperature period, irrespective of the male. Similarly, some cytokines showed differences between daylength/temperature periods when focusing on their total amount in the ejaculate. No strong relationship (explaining more than 50% of the total variance) was found between annual fluctuations in SP-cytokine levels and semen parameters. In conclusion, the period of the year during which ejaculates were collected helps explain the intra-male variability of SP-cytokine levels in breeding boars.
Product Description
Millipore
MILLIPLEX® Porcine Cytokine/Chemokine Magnetic Bead Panel - Immunology Multiplex Assay, Simultaneously analyze multiple cytokine and chemokine biomarkers with Bead-Based Multiplex Assays using the Luminex technology, in porcine serum, plasma and cell culture samples.