MilliporeSigma
Few-Layer Graphene-Based Nanofluids with Enhanced Thermal Conductivity.
Nanomaterials (Basel, Switzerland) (2020-07-02)
Samah Hamze, Nawal Berrada, David Cabaleiro, Alexandre Desforges, Jaafar Ghanbaja, Jérôme Gleize, Dominique Bégin, Florentin Michaux, Thierry Maré, Brigitte Vigolo, Patrice Estellé
ABSTRACT
High-quality graphene is an especially promising carbon nanomaterial for developing nanofluids for enhancing heat transfer in fluid circulation systems. We report a complete study on few layer graphene (FLG) based nanofluids, including FLG synthesis, FLG-based nanofluid preparation, and their thermal conductivity. The FLG sample is synthesized by an original mechanical exfoliation method. The morphological and structural characterization are investigated by both scanning and transmission electron microscopy and Raman spectroscopy. The chosen two-step method involves the use of thee nonionic surfactants (Triton X-100, Pluronic® P123, and Gum Arabic), a commercial mixture of water and propylene glycol and a mass content in FLG from 0.05 to 0.5%. The thermal conductivity measurements of the three FLG-based nanofluid series are carried out in the temperature range 283.15-323.15 K by the transient hot-wire method. From a modeling analysis of the nanofluid thermal conductivity behavior, it is finally shown that synergetic effects of FLG nanosheet size and thermal resistance at the FLG interface both have significant impact on the evidenced thermal conductivity enhancement.
Product Description
Supelco
Dioctyl phenylphosphonate, Selectophore
Sigma-Aldrich
Triton X-100, laboratory grade