MilliporeSigma
  • Timing of appearance of new mutations during yeast meiosis and their association with recombination.
Timing of appearance of new mutations during yeast meiosis and their association with recombination.
Current genetics (2020-01-15)
Osama Mansour, Liat Morciano, Keren Zion, Renana Elgrabli, Drora Zenvirth, Giora Simchen, Ayelet Arbel-Eden
ABSTRACT
Mutations in budding yeast occur in meiosis at higher frequencies than in cells grown vegetatively. In contrast to mutations that occur in somatic cells, meiotic mutations have a special, long-range impact on evolution, because they are transferred to the following generations through the gametes. Understanding the mechanistic basis of meiotic mutagenicity is still lacking, however. Here, we report studies of mutagenicity in the reporter gene CAN1, in which forward mutation events in meiosis are sevenfold higher than in mitotic cells, as determined by fluctuation analysis. Meiotic mutations appear approximately at the same time as heteroallelic-recombination products and as meiotic DSBs. Recombination-associated timing of meiotic mutagenicity is further augmented by the absence of meiotic mutations in cells arrested after pre-meiotic DNA synthesis. More than 40% of the mutations generated in meiosis in CAN1 are found on chromosomes that have recombined in the 2.2 kb covering the reporter, implying that the mutations have resulted from recombination events and that meiotic recombination is mutagenic. The induced expression in yeast meiosis of low-fidelity DNA polymerases coded by the genes REV1, REV3, RAD30, and POL4 makes them attractive candidates for introducing mutations. However, in our extensive experiments with polymerase-deleted strains, these polymerases do not appear to be the major source of meiotic mutagenicity. From the connection between meiotic mutagenicity and recombination, one may conclude that meiotic recombination has another diversification role, of introducing new mutations at the DNA sequence level, in addition to reshuffling of existing variation. The new, rare meiotic mutations may contribute to long-range evolutionary processes and enhance adaptation to challenging environments.
Sigma-Aldrich
L-Canavanine sulfate salt, ≥99% (TLC), powder