MilliporeSigma
  • Tetrabutylammonium tosylate as inert phase-transfer catalyst: The key to high efficiency SN2 radiofluorinations.
Tetrabutylammonium tosylate as inert phase-transfer catalyst: The key to high efficiency SN2 radiofluorinations.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine (2020-06-21)
Viktoriya Orlovskaya, Dmitrii Antuganov, Olga Fedorova, Vasilii Timofeev, Raisa Krasikova
ABSTRACT
Here we describe use of the tetrabutylammonium p-toluenesulfonate (TBAOTs) as an inert phase transfer catalyst allowing high efficient aliphatic radiofluorination of labeling precursors for the preparation of common radiotracers using only 0.2-1.2 mg of the substrates. With operational convenience (no need for azeotropic drying) and compatibility with the common types of strong anion-exchange cartridges (QMA), this method has a high potential for routine application in various automated synthesizers and offers significant costs reductions due to less precursors used.
Sigma-Aldrich
1-(3-Hydroxypropyl)-1H-imidazole