MilliporeSigma
  • Fast deposition of an ultrathin, highly crystalline organic semiconductor film for high-performance transistors.
Fast deposition of an ultrathin, highly crystalline organic semiconductor film for high-performance transistors.
Nanoscale horizons (2020-05-20)
Xiali Zhang, Wei Deng, Bei Lu, Xiaochen Fang, Xiujuan Zhang, Jiansheng Jie
ABSTRACT
Ultrathin organic semiconductor (OSC) crystalline films hold the promise of achieving high-performance, flexible, and transparent organic electronic devices. However, fast and high-throughput solution deposition of uniform pinhole-free ultrathin OSC crystalline films over a large area remains a challenge. Here, we demonstrate that a mixed solvent system can obviously alter the fluid flow dynamics and significantly improve the blade-coating quality of the film, enabling us to achieve a large-area continuous and smooth bis(triethylsilylethynyl)anthradithiophene (Dif-TES-ADT) ultrathin film at a fast coating speed of ∼1 mm s-1, much superior to the 30-50 μm s-1 for conventional methods. Also, the ultrathin, highly crystalline Dif-TES-ADT film-based organic thin-film transistors (OTFTs) exhibit a maximum mobility up to 5.54 cm2 V-1 s-1, which is on par with the Dif-TES-ADT single crystal-based devices and among the highest for Dif-TES-ADT film-based devices. This finding should open a new route to achieve ultrathin OSC crystalline film-based high-performance flexible and transparent electronics.
Sigma-Aldrich
N,N′-Dioctyl-3,4,9,10-perylenedicarboximide, 98%