MilliporeSigma
Organocatalyzed Birch Reduction Driven by Visible Light.
Journal of the American Chemical Society (2020-07-15)
Justin P Cole, Dian-Feng Chen, Max Kudisch, Ryan M Pearson, Chern-Hooi Lim, Garret M Miyake
ABSTRACT
The Birch reduction is a powerful synthetic methodology that uses solvated electrons to convert inert arenes to 1,4-cyclohexadienes-valuable intermediates for building molecular complexity. Birch reductions traditionally employ alkali metals dissolved in ammonia to produce a solvated electron for the reduction of unactivated arenes such as benzene (Ered < -3.42 V vs SCE). Photoredox catalysts have been gaining popularity in highly reducing applications, but none have been reported to demonstrate reduction potentials powerful enough to reduce benzene. Here, we introduce benzo[ghi]perylene imides as new organic photoredox catalysts for Birch reductions performed at ambient temperature and driven by visible light from commercially available LEDs. Using low catalyst loadings (<1 mol percent), benzene and other functionalized arenes were selectively transformed to 1,4-cyclohexadienes in moderate to good yields in a completely metal-free reaction. Mechanistic studies support that this unprecedented visible-light-induced reactivity is enabled by the ability of the organic photoredox catalyst to harness the energy from two visible-light photons to affect a single, high-energy chemical transformation.
Sigma-Aldrich
Isochroman, 99%