MilliporeSigma
An ancient push-pull pollination mechanism in cycads.
Science advances (2020-06-26)
Shayla Salzman, Damon Crook, James D Crall, Robin Hopkins, Naomi E Pierce
ABSTRACT
Most cycads engage in brood-site pollination mutualisms, yet the mechanism by which the Cycadales entice pollination services from diverse insect mutualists remains unknown. Here, we characterize a push-pull pollination mechanism between a New World cycad and its weevil pollinators that mirrors the mechanism between a distantly related Old World cycad and its thrips pollinators. The behavioral convergence between weevils and thrips, combined with molecular phylogenetic dating and a meta-analysis of thermogenesis and coordinated patterns of volatile attraction and repulsion suggest that a push-pull pollination mutualism strategy is ancestral in this ancient, dioecious plant group. Hence, it may represent one of the earliest insect/plant pollination mechanisms, arising long before the evolution of visual floral signaling commonly used by flowering plants.
Sigma-Aldrich
Dichloromethane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Supelco
Glass Wool, Silanized, pkg of 50 g
Porapak Porous Polymer Adsorbent, matrix Porapak Q, 80-100 mesh, bottle of 75 cc