MilliporeSigma
Lipase 8 affects the pathogenesis of Candida albicans.
Infection and immunity (2007-07-25)
Attila Gácser, Frank Stehr, Cathrin Kröger, László Kredics, Wilhelm Schäfer, Joshua D Nosanchuk
ABSTRACT
The production of lipases can affect microbial fitness and virulence. We examined the role of the lipase 8 (LIP8) gene in the virulence of Candida albicans by constructing Deltalip8 strains by the URA-blaster disruption method. Reverse transcription-PCR experiments demonstrated the absence of LIP8 expression in the homozygous knockout mutants. Reconstituted strains and overexpression mutants were generated by introducing a LIP8 open reading frame under control of a constitutive actin promoter. Knockout mutants produced more mycelium, particularly at higher temperatures and pH >or=7. Diminished LIP8 expression resulted in reduced growth in lipid-containing media. Mutants deficient in the LIP8 gene were significantly less virulent in a murine intravenous infection model. The results clearly indicate that Lip8p is an important virulence factor of C. albicans.
Sigma-Aldrich
Lipase from Candida sp., recombinant, expressed in Aspergillus niger