MilliporeSigma
  • Species differences in the formation of DA-8164 after intravenous and/or oral administration of DA-8159, a new erectogenic, to mice, rats, rabbits, dogs and humans.
Species differences in the formation of DA-8164 after intravenous and/or oral administration of DA-8159, a new erectogenic, to mice, rats, rabbits, dogs and humans.
Biopharmaceutics & drug disposition (2005-04-02)
Hyun J Shim, Yu C Kim, Joo H Lee, Kyung J Park, Jong W Kwon, Won B Kim, Myung G Lee
ABSTRACT
Species differences in the formation of DA-8164 after intravenous and/or oral administration of DA-8159 to mice, rats, rabbits, dogs and humans were investigated. After intravenous administration of DA-8159, the formation of DA-8164 decreased in the order mice, rats, rabbits and dogs; the AUC(DA-8164)/AUC(DA-8159) ratios were 0.479, 0.199, 0.0452 and close to 0 (DA-8164 was below the detection limit in dog plasma), respectively. After oral administration of DA-8159, the formation of DA-8164 was considerable in mice, rats and humans, but almost negligible in dogs; the AUC (or AUC(0-t))(DA-8164)/AUC (or AUC(0-t))(DA-8159) ratios were 2.99, 2.67, 1.39 and 0.0650, respectively. The above data suggested that the formation of DA-8164 was almost negligible after both intravenous and oral administration in dogs. The species differences for the formation of DA-8164 may be due to the involvement of different CYP isozymes for each species and/or a different amount or activity of CYP isozyme if the same CYP isozyme is involved for the formation of DA-8164 for all species. The AUC (or AUC(0-t))(DA-8164)/AUC (or AUC(0-t))(DA-8159) ratios after oral administration were greater than those after intravenous administration in mice, rats and dogs, and this could be due to considerable first-pass (gastric, intestinal and/or hepatic) effects in the species as proved in rats.
Sigma-Aldrich
Udenafil, ≥98% (HPLC)