MilliporeSigma
  • MUC1 expression in papillary thyroid carcinoma is associated with BRAF mutation and lymph node metastasis; the latter is the most important risk factor of relapse.
MUC1 expression in papillary thyroid carcinoma is associated with BRAF mutation and lymph node metastasis; the latter is the most important risk factor of relapse.
Thyroid : official journal of the American Thyroid Association (2014-07-12)
Florence Renaud, Viviane Gnemmi, Patrick Devos, Sébastien Aubert, Michel Crépin, Lucie Coppin, Nassima Ramdane, Brigitte Bouchindhomme, Michelle d'Herbomez, Isabelle Van Seuningen, Christine Do Cao, François Pattou, Bruno Carnaille, Pascal Pigny, Jean-Louis Wémeau, Emmanuelle Leteurtre
ABSTRACT
The incidence of papillary thyroid carcinoma (PTC) has increased over the past 30 years in Western countries. PTC is usually associated with a good prognosis, but there is a wide range of aggressiveness, and some patients develop distant metastasis and/or resistance to standard treatment. Early identification of these high-risk tumors is a current challenge for appropriate patient management. MUC1 expression has been studied previously in thyroid cancer, but its prognostic value remains controversial. Here, we correlated MUC1 expression in PTC with clinical and pathological features and with the presence of the BRAF(V600E) mutation. We performed a clinical and morphological analysis of 190 thyroid tumors (95 PTCs and 95 adenomas). MUC1 immunohistochemistry was carried out on a tissue microarray using different antibodies. The presence of the BRAF(V600E) mutation was investigated by pyrosequencing. MUC1 mRNA levels were assessed by quantitative reverse transcription polymerase chain reaction on a subset of PTC. MUC1 expression was observed in 49% of PTCs and was found to correlate with the presence of papillary architecture, a stromal lymphoid infiltrate, aggressive histological subtypes, extrathyroidal extension, lymph node metastasis, nuclear pseudoinclusions, lymphovascular invasion, and the presence of the BRAF(V600E) mutation (p<0.0001). MUC1 was abundant in nuclear pseudoinclusions. Multivariate analysis showed a strong association of MUC1 expression with the presence of the BRAF(V600E) mutation and lymph node metastasis (p<0.0001). Lymph node metastasis was the most important risk factor of relapse. Our study shows an association between MUC1 expression and the presence of the BRAF(V600E) mutation in PTC. Analysis of MUC1 expression could improve the risk stratification of PTCs.
Sigma-Aldrich
MUC1 (EP85) Rabbit Monoclonal Primary Antibody