MilliporeSigma
  • The stress polarity signaling (SPS) pathway serves as a marker and a target in the leaky gut barrier: implications in aging and cancer.
The stress polarity signaling (SPS) pathway serves as a marker and a target in the leaky gut barrier: implications in aging and cancer.
Life science alliance (2020-02-12)
Pradipta Ghosh, Lee Swanson, Ibrahim M Sayed, Yash Mittal, Blaze B Lim, Stella-Rita Ibeawuchi, Marc Foretz, Benoit Viollet, Debashis Sahoo, Soumita Das
ABSTRACT
The gut barrier separates trillions of microbes from the largest immune system in the body; when compromised, a "leaky" gut barrier fuels systemic inflammation, which hastens the progression of chronic diseases. Strategies to detect and repair the leaky gut barrier remain urgent and unmet needs. Recently, a stress-polarity signaling (SPS) pathway has been described in which the metabolic sensor, AMP-kinase acts via its effector, GIV (also known as Girdin) to augment epithelial polarity exclusively under energetic stress and suppresses tumor formation. Using murine and human colon-derived organoids, and enteroid-derived monolayers (EDMs) that are exposed to stressors, we reveal that the SPS-pathway is active in the intestinal epithelium and requires a catalytically active AMP-kinase. Its pharmacologic augmentation resists stress-induced collapse of the epithelium when challenged with microbes or microbial products. In addition, the SPS-pathway is suppressed in the aging gut, and its reactivation in enteroid-derived monolayers reverses aging-associated inflammation and loss of barrier function. It is also silenced during progression of colorectal cancers. These findings reveal the importance of the SPS-pathway in the gut and highlights its therapeutic potential for treating gut barrier dysfunction in aging, cancer, and dysbiosis.
Sigma-Aldrich
Anti-Girdin, coiled-coil region Antibody, from rabbit, purified by affinity chromatography