MilliporeSigma
  • Cooperative dual catalysis: application to the highly enantioselective conjugate cyanation of unsaturated imides.
Cooperative dual catalysis: application to the highly enantioselective conjugate cyanation of unsaturated imides.
Journal of the American Chemical Society (2004-08-12)
Glenn M Sammis, Hiroshi Danjo, Eric N Jacobsen
ABSTRACT
Cooperative heterobimetallic catalysis was used as a design principle to achieve a highly reactive system for the enantioselective conjugate addition of cyanide to alpha,beta-unsaturated imides. A dual-catalyst pathway involving chiral (salen)Al complex 1b and chiral (pybox)Er complex 4b provides measurable improvements in rates and enantioselectivities relative to single-catalyst systems. Mechanistic studies point to a cooperative bimetallic mechanism involving activation of the imide by the Al complex and activation of cyanide by the Er complex.
Sigma-Aldrich
(S,S)-N,N′-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminoaluminum chloride