MilliporeSigma
  • Effects of GLP-1-(7-36)NH2, GLP-1-(7-37), and GLP-1- (9-36)NH2 on intravenous glucose tolerance and glucose-induced insulin secretion in healthy humans.
Effects of GLP-1-(7-36)NH2, GLP-1-(7-37), and GLP-1- (9-36)NH2 on intravenous glucose tolerance and glucose-induced insulin secretion in healthy humans.
The Journal of clinical endocrinology and metabolism (2003-04-08)
Torsten P Vahl, Breay W Paty, Bradley D Fuller, Ronald L Prigeon, David A D'Alessio
ABSTRACT
Glucagon-like peptide 1 (GLP-1) is an insulin secretagogue synthesized in the intestine and released in response to meal ingestion. It is secreted primarily in two forms, GLP-1-(7-37) and GLP-1-(7-36)NH(2), both of which bind to a specific GLP-1 receptor (GLP-1r) on the pancreatic beta-cell and augment glucose-stimulated insulin secretion. Once secreted, GLP-1-(7-36)NH(2) is rapidly metabolized to GLP-1-(9-36)NH(2), which is the predominant form of GLP-1 in postprandial plasma because of its relatively slower clearance. Although no clear biological role for GLP-1-(9-36)NH(2) in humans has been identified, recent studies in animals suggest two potential effects: to antagonize the effects of intact GLP-1 and to promote glucose disappearance in peripheral tissues. In the studies reported here we compared the independent effects of GLP-1-(7-36)NH(2), GLP-1-(7-37), and GLP-1-(9-36)NH(2) on parameters of iv glucose tolerance and determined whether GLP-1-(9-36)NH(2) inhibits the insulinotropic actions of GLP-1. Ten healthy subjects underwent 4 separate frequently sampled iv glucose tolerance tests during infusions of GLP-1-(7-37), GLP-1-(7-36)NH(2), GLP-1-(9-36)NH(2), or saline. Results from the iv glucose tolerance test were used to obtain indexes of beta-cell function (acute insulin response to glucose) and iv glucose tolerance (glucose disappearance constant), and the minimal model of glucose kinetics was used to obtain indexes of glucose effectiveness and insulin sensitivity. Compared with control studies, both GLP-1-(7-36)NH(2) and GLP-1-(7-37) significantly increased acute insulin response to glucose, glucose disappearance constant, glucose effectiveness, and glucose effectiveness at zero insulin, but did not change the insulin sensitivity index. In contrast, none of the parameters of glucose tolerance was measurably affected by GLP-1-(9-36) amide. In a second set of experiments, 10 healthy subjects had glucose-stimulated insulin secretion measured during an infusion of GLP-1-(7-36)NH(2) alone or with a simultaneous infusion of GLP-1-(9-36)NH(2) that increased plasma levels approximately 10-fold over those produced by unmetabolized GLP-1. Augmentation of glucose-stimulated insulin secretion by GLP-1-(7-36)NH(2) was not altered by the coadministration of GLP-1-(9-36)NH(2). Based on these results we conclude that GLP-1-(9-36)NH(2) does not regulate insulin release or glucose metabolism in healthy humans.
Sigma-Aldrich
Glucagon-Like Peptide 1 Fragment 7-37 human, ≥96% (HPLC)