MilliporeSigma
  • cis-4-Methylsphingosine decreases sphingolipid biosynthesis by specifically interfering with serine palmitoyltransferase activity in primary cultured neurons.
cis-4-Methylsphingosine decreases sphingolipid biosynthesis by specifically interfering with serine palmitoyltransferase activity in primary cultured neurons.
The Journal of biological chemistry (1997-06-20)
G van Echten-Deckert, A Zschoche, T Bär, R R Schmidt, A Raths, T Heinemann, K Sandhoff
ABSTRACT
The effect of six different structurally modified sphingosine analogues on biosynthesis of sphingolipids was studied in primary cultured murine cerebellar neurons. Treatment of cells with cis-4-methylsphingosine at micromolar levels resulted in a markedly decreased sphingolipid biosynthesis, whereas the other compounds examined, trans-4-methylsphingosine, cis-5-methylsphingosine, trans-5-methylsphingosine, cis-sphingosine, and 1-deoxysphingosine, inhibited sphingolipid biosynthesis less efficiently. The inhibition of sphingolipid biosynthesis by the various compounds was paralleled by a decrease of serine palmitoyltransferase activity in situ. For cis-4-methylsphingosine the inhibitory effect on serine palmitoyltransferase activity was shown to be concentration- and time-dependent. Half-maximal reduction of enzyme activity occurred after 24 h of treatment with 10 microM of the compound. The activity of other enzymes of sphingolipid biosynthesis as well as phospholipid and protein biosynthesis was not affected. Analysis of the sphingoid moiety of cellular sphingolipids suggests that the sphingosine analogues listed above were subject to degradation rather than being utilized as precursors for sphingolipid biosynthesis by cultured neurons. Except of 1-deoxysphingosine, the other five sphingosine analogues were shown to be substrates for sphingosine kinase in vitro. After 24 h of treatment of primary cerebellar neurons with the various sphingosine analogues the relative percentage of the respective intracellular 1-phosphate derivatives paralleled exactly the inhibitory effect on serine palmitoyltransferase activity observed when cells were treated with the unphosphorylated compounds. In contrast to the respective 1-phosphate derivatives of the other methyl-branched sphingosine analogues examined, cis-4-methylsphingosine 1-phosphate showed an intracellular accumulation suggesting a delayed turnover rate in cultured murine neurons for this compound. These results suggest that the inhibitory effect of the sphingosine analogues on serine palmitoyltransferase is mediated by their respective 1-phosphate derivatives and that the pronounced effect of cis-4-methylsphingosine is caused by a high intracellular concentration of cis-4-methylsphingosine 1-phosphate. cis-4-Methylsphingosine, in addition, caused drastic changes in cell morphology of primary cerebellar neurons, which were not observed when these cells were treated with one of the other sphingosine analogues examined.
Product Description
Avanti
cis-4-methylsphingosine, Avanti Research - A Croda Brand
Avanti
16:0-d31-18:1 PI, 1-palmitoyl-d31-2-oleoyl-sn-glycero-3-phosphoinositol (ammonium salt), powder