MilliporeSigma
  • The auxin-induced maize gene ZmSAUR2 encodes a short-lived nuclear protein expressed in elongating tissues.
The auxin-induced maize gene ZmSAUR2 encodes a short-lived nuclear protein expressed in elongating tissues.
The Journal of biological chemistry (2003-04-16)
Stefan Knauss, Thomas Rohrmeier, Ludwig Lehle
ABSTRACT
By differential screening of a cDNA library from auxin-induced maize coleoptiles we have isolated and characterized a SAUR gene, designated ZmSAUR2, belonging to a not yet characterized subtype of the SAUR family. ZmSAUR2 encodes a 15.3-kDa protein and is specifically induced by auxin in elongating coleoptile tissue but not in primary leaves or in roots. The transcript level rapidly increased within minutes and preceded auxin-stimulated elongation of coleoptile segments. Cycloheximide also induced ZmSAUR2 transcription, as has been shown for other early auxin-induced genes, whereas abscisic acid, brassinolide, ethylene, gibberellic acid, kinetin, and methyl jasmonate did not provoke an increase in ZmSAUR2 mRNA abundance. In pulse-chase experiments using auxin-induced coleoptiles and an anti-ZmSAUR2 antibody we were able to precipitate a protein of the expected molecular mass and to determine a half-life of about 7 min, which is among the shortest known in eukaryotes. In gel shift assays binding of calmodulin to ZmSAUR2 was demonstrated, suggesting the possibility of post-transcriptional regulation. Upon transformation of onion epidermal cells with a ZmSAUR2::GUS construct the corresponding chimeric protein was detected in the nucleus. The results suggest that ZmSAUR2 encodes a short-lived nuclear protein that might be involved in auxin-mediated cell elongation.
Sigma-Aldrich
Chu (N6) Basal Salt Mixture, powder, suitable for plant cell culture