Merck
  • Structure-Activity Relationship for Small Molecule Inhibitors of Nicotinamide N-Methyltransferase.
Structure-Activity Relationship for Small Molecule Inhibitors of Nicotinamide N-Methyltransferase.
Journal of medicinal chemistry (2017-05-27)
Harshini Neelakantan, Hua-Yu Wang, Virginia Vance, Jonathan D Hommel, Stanton F McHardy, Stanley J Watowich
ABSTRACT
Nicotinamide N-methyltransferase (NNMT) is a fundamental cytosolic biotransforming enzyme that catalyzes the N-methylation of endogenous and exogenous xenobiotics. We have identified small molecule inhibitors of NNMT with >1000-fold range of activity and developed comprehensive structure-activity relationships (SARs) for NNMT inhibitors. Screening of N-methylated quinolinium, isoquinolinium, pyrididium, and benzimidazolium/benzothiazolium analogues resulted in the identification of quinoliniums as a promising scaffold with very low micromolar (IC50 ∼ 1 μM) NNMT inhibition. Computer-based docking of inhibitors to the NNMT substrate (nicotinamide)-binding site produced a robust correlation between ligand-enzyme interaction docking scores and experimentally calculated IC50 values. Predicted binding orientation of the quinolinium analogues revealed selective binding to the NNMT substrate-binding site residues and essential chemical features driving protein-ligand intermolecular interactions and NNMT inhibition. The development of this new series of small molecule NNMT inhibitors direct the future design of lead drug-like inhibitors to treat several metabolic and chronic disease conditions characterized by abnormal NNMT activity.
Sigma-Aldrich
5-Amino-1-methylquinolinium iodide, ≥98% (HPLC)