MilliporeSigma
  • mRNA guanylation catalyzed by the S-adenosylmethionine-dependent guanylyltransferase of bamboo mosaic virus.
mRNA guanylation catalyzed by the S-adenosylmethionine-dependent guanylyltransferase of bamboo mosaic virus.
The Journal of biological chemistry (2005-01-29)
Yih-Leh Huang, Yau-Heiu Hsu, Yu-Tsung Han, Menghsiao Meng
ABSTRACT
The S-adenosylmethionine-dependent guanylyltransferase of bamboo mosaic virus belongs to a novel class of mRNA capping enzymes distantly conserved in Alphavirus-like superfamily. The reaction sequence of the viral enzyme has been proposed comprising steps of 1) binding of GTP and S-adenosylmethionine, 2) formation of m7GTP and S-adenosylhomocysteine, 3) formation of the covalent (Enzyme-m7GMP) intermediate, and 4) transfer of m7GMP from the intermediate to the RNA acceptor. In this study the acceptor specificity of the viral enzyme was characterized. The results show that adenylate or guanylate with 5'-diphosphate group is an essential feature for acceptors, which can be RNA or mononucleotide, to receive m7GMP. The transfer rate of m7GMP to guanylate is greater than to adenylate by a factor of approximately 3, and the K(m) value for mononucleotide acceptor is approximately 10(3)-fold higher than that for RNA. The capping efficiency of the viral genomic RNA transcript depends on the length of the transcript and the formation of a putative stem-loop structure, suggesting that mRNA capping process may participate in regulating the viral gene expression.
Sigma-Aldrich
7-Methylguanosine 5′-triphosphate sodium salt, ≥85% (HPLC)