MilliporeSigma
  • Interactions of alamethicin with model cell membranes investigated using sum frequency generation vibrational spectroscopy in real time in situ.
Interactions of alamethicin with model cell membranes investigated using sum frequency generation vibrational spectroscopy in real time in situ.
The journal of physical chemistry. B (2010-02-19)
Shuji Ye, Khoi Tan Nguyen, Zhan Chen
ABSTRACT
Structures of membrane-associated peptides and molecular interactions between peptides and cell membrane bilayers govern biological functions of these peptides. Sum frequency generation (SFG) vibrational spectroscopy has been demonstrated to be a powerful technique to study such structures and interactions at the molecular level. In this research, SFG has been applied, supplemented by attenuated total reflectance Fourier transform infrared spectroscopy (ATR-FTIR), to characterize the interactions between alamethicin (a model for larger channel proteins) and different lipid bilayers in the absence of membrane potential. The orientation of alamethicin in lipid bilayers has been determined using SFG amide I spectra detected with different polarization combinations. It was found that alamethicin adopts a mixed alpha-helical and 3(10)-helical structure in fluid-phase lipid bilayers. The helix (mainly alpha-helix) at the N-terminus tilts at about 63 degrees versus the surface normal in a fluid-phase 1,2-dimyristoyl-d54-sn-glycero-3-phosphocholine-1,1,2,2-d4-N,N,N-trimethyl-d9 (d-DMPC)/1,2-dimyristoyl-sn-glycero-3-phosphocholine (DMPC) bilayer. The 3(10)-helix at the C-terminus (beyond the Pro14 residue) tilts at about 43 degrees versus the surface normal. This is the first time to apply SFG to study a 3(10)-helix experimentally. When interacting with a gel-phase lipid bilayer, alamethicin lies down on the gel-phase bilayer surface or aggregates or both, which does not have significant insertion into the lipid bilayer.
Product Description
Avanti
16:0 PG-d62, Avanti Research - A Croda Brand 860382P, powder