MilliporeSigma
  • PPARgamma1-induced caveolin-1 enhances cholesterol efflux and attenuates atherosclerosis in apolipoprotein E-deficient mice.
PPARgamma1-induced caveolin-1 enhances cholesterol efflux and attenuates atherosclerosis in apolipoprotein E-deficient mice.
Journal of vascular research (2009-09-05)
Qin Hu, Xian-Jun Zhang, Chun-Xi Liu, Xu-Ping Wang, Yun Zhang
ABSTRACT
Caveolin-1 (Cav-1) may positively or negatively influence the development of atherosclerosis, depending on the cell type and the metabolic pathways regulated by this protein. We investigate the effects of Cav-1 on cholesterol efflux in RAW264.7 infected with AdPPARgamma1 and whether Cav-1 could attenuate established atherosclerotic lesions in PPARgamma1-treated apoE-deficient mice. Compared with AdGFP control, PPARgamma1 and Cav-1 were constitutively overexpressed in AdPPARgamma1-infected RAW264.7 cells, which stimulated cholesterol efflux to apolipoprotein A-I. Using a small interfering RNA approach (Cav-1-siRNA) we achieved an efficient and specific knockdown of caveolin-1 expression (80%), which resulted in a remarkable reduction of cholesterol efflux in RAW264.7 cells . Moreover, PPARgamma1-treated Cav-1-siRNA RAW264.7 cells showed more ability to stimulate cholesterol efflux than Cav-1-siRNA RAW264.7 cells, but far less than control-siRNA RAW264.7 cells and PPARgamma1-treated RAW264.7 cells. In addition, 40-week-old apoE-deficient mice fed a Western-type diet and infected for 4 weeks with AdPPARgamma1 showed induced Cav-1 expression in aortic vascular endothelial cells, smooth muscle cells and macrophages, as well as attenuated established atherosclerotic lesions. PPARgamma1 gene therapy could induce Cav-1 expression and enhance cholesterol efflux and attenuate atherosclerosis in apoE-deficient mice.
Sigma-Aldrich
Anti-Macrophages/Monocytes Antibody, clone MOMA-2, clone MOMA-2, Chemicon®, from rat