MilliporeSigma
  • S-(4-Methoxyphenyl)-4-methoxybenzenesulfonothioate as a Promising Lead Compound for the Development of a Renal Carcinoma Agent.
S-(4-Methoxyphenyl)-4-methoxybenzenesulfonothioate as a Promising Lead Compound for the Development of a Renal Carcinoma Agent.
ChemMedChem (2019-12-14)
Camilla I Nantes, Ingrid D Pereira, Ruoli Bai, Ernest Hamel, James C Burnett, Rodrigo J de Oliveira, Maria de F C Matos, Adilson Beatriz, Murilo K A Yonekawa, Renata T Perdomo, Dênis P de Lima, Danielle Bogo, Edson Dos A Dos Santos
ABSTRACT
Organosulfur compounds show cytotoxic potential towards many tumor cell lines. Disulfides and thiosulfonates act through apoptotic processes, inducing proteins associated with apoptosis, endoplasmic reticulum stress, and the unfolded protein response. Three p-substituted symmetric diaryl disulfides and three diaryl thiosulfonates were synthesized and analyzed for inhibition of tubulin polymerization and for human cancer cell cytotoxic activity against seven tumor cell lines and a non-tumor cell line. S-(4-methoxyphenyl)-4-methoxybenzenesulfonothioate (6) exhibited inhibition of tubulin polymerization and showed the best antiproliferative potential, especially against the 786-0 cell line, being six times more selective as compared with the non-tumor cell line. In addition, compound 6 was able to activate caspase-3 after 24 and 48 h treatments of the 786-0 cell line and induced cell-cycle arrest in the G2/M stage at the highest concentration evaluated at 24 and 48 h. Compound 6 was able to cause complete inhibition of proliferation, inducing the death of 786-0 cells, by increasing the number of cells at G2/M and greater activation of caspase-3.
Sigma-Aldrich
4-Methylbenzenethiol, 98%
Sigma-Aldrich
7-Aminoactinomycin D, ~97% (HPLC), powder