MilliporeSigma
  • Inhibition of Swarming of Proteus by Sodium Tetradecyl Sulfate, beta-Phenethyl Alcohol, and p-Nitrophenylglycerine.
Inhibition of Swarming of Proteus by Sodium Tetradecyl Sulfate, beta-Phenethyl Alcohol, and p-Nitrophenylglycerine.
Applied microbiology (1966-11-01)
R Kopp, J Müller, R Lemme
ABSTRACT
The effects of sodium tetradecyl sulfate (STS), beta-phenethyl alcohol (PEA), and p-nitrophenylglycerol (PNPG) on motility, swarming, flagellation, and growth of Proteus were examined. Growth-inhibitory concentrations (GIC) and swarming-inhibitory concentrations (SIC) were determined. A characterization of the swarming-inhibitory efficacy of these compounds was based on their GIC/SIC ratio and their concentration inhibition curves. Using the homologous series of sodium alkyl sulfates as a standard reference, we showed that PNPG was more effective than STS, which was the most effective of the homologous series. PEA was less effective than sodium decyl sulfate but more effective than sodium octyl sulfate. Motility tests in liquid medium and electron microscope investigations indicated that the modes of action of the three compounds, all of which effectively inhibit the swarming of Proteus, are different. Whereas STS and PEA inhibit swarming by inhibition of motility, PNPG seems to act on the swarming mechanism sensu strictori, without impairment of motility. STS immobilizes by inhibition of flagellum formation or by some lytic action on the flagella already synthesized. PEA acts by impairing flagellar function, but leaves the flagella morphologically intact.
Sigma-Aldrich
1-(4-Nitrophenyl)glycerol, crystalline