MilliporeSigma
  • α-synuclein Induces Mitochondrial Dysfunction through Spectrin and the Actin Cytoskeleton.
α-synuclein Induces Mitochondrial Dysfunction through Spectrin and the Actin Cytoskeleton.
Neuron (2017-12-19)
Dalila G Ordonez, Michael K Lee, Mel B Feany
ABSTRACT
Genetics and neuropathology strongly link α-synuclein aggregation and neurotoxicity to the pathogenesis of Parkinson's disease and related α-synucleinopathies. Here we describe a new Drosophila model of α-synucleinopathy based on widespread expression of wild-type human α-synuclein, which shows robust neurodegeneration, early-onset locomotor deficits, and abundant α-synuclein aggregation. We use results of forward genetic screening and genetic analysis in our new model to demonstrate that α-synuclein expression promotes reorganization of the actin filament network and consequent mitochondrial dysfunction through altered Drp1 localization. Similar changes are present in a mouse α-synucleinopathy model and in postmortem brain tissue from patients with α-synucleinopathy. Importantly, we provide evidence that the interaction of α-synuclein with spectrin initiates pathological alteration of the actin cytoskeleton and downstream neurotoxicity. These findings suggest new therapeutic approaches for α-synuclein induced neurodegeneration.
Sigma-Aldrich
Anti-Aggregated a-Synuclein Antibody, clone 5G4, clone 5G4, from mouse, purified by affinity chromatography