MilliporeSigma
  • Dioscin inhibits stem-cell-like properties and tumor growth of osteosarcoma through Akt/GSK3/β-catenin signaling pathway.
Dioscin inhibits stem-cell-like properties and tumor growth of osteosarcoma through Akt/GSK3/β-catenin signaling pathway.
Cell death & disease (2018-03-03)
Weihai Liu, Zhiqiang Zhao, Yongqian Wang, Wuguo Li, Qiao Su, Qiang Jia, Jiajun Zhang, Xuelin Zhang, Jingnan Shen, Junqiang Yin
ABSTRACT
Osteosarcoma is the most common primary bone tumor in children and adolescents. Many patients with osteosarcoma always develop drug resistance to current chemotherapy regimens, which induces a poor prognosis. And cancer stem cells (CSCs) have been reported to possess the properties to self-renew and maintain the phenotype of tumor, which may lead to clinical treatment failure. Thus, it is an urgent task to develop several potentially useful therapeutic agents, which could target CSCs in osteosarcoma. This study aims to clarify the in vitro and in vivo anti-osteosarcoma effects of dioscin, the primary component derived from Discorea nipponica Makino, and its molecular mechanism of action. In this study, all the ten human osteosarcoma cell lines were sensitive to dioscin treatment in a dose- and time-dependent manner. Dioscin inhibits proliferation and induces cell cycle arrest as well as apoptotic cell death in osteosarcoma cells. More importantly, oral administration of dioscin (60 mg/kg) showed significant therapeutic effect on osteosarcoma growth without obvious side effects in vivo. In addition, dioscin possesses the ability to suppress stem-cell-like phenotype of osteosarcoma cells. Mechanistically, dioscin inhibits osteosarcoma stem-cell-like properties and tumor growth through repression of Akt/GSK3/β-catenin pathway. Moreover, β-catenin expression in osteosarcoma patients was associated with clinical prognosis. Conclusively, the present study provides comprehensive evidence for the inhibition of dioscin on osteosarcoma stem-cell-like properties and tumor growth through repression of Akt/GSK3/β-catenin pathway, which suggests dioscin as a promising therapeutic regimen. And β-catenin may be a potential therapeutic target as well as a significant prognostic marker for osteosarcoma patients in clinic.
Sigma-Aldrich
Anti-Notch 1 Antibody, NT, serum, from rabbit