MilliporeSigma
A LCMT1-PME-1 methylation equilibrium controls mitotic spindle size.
Cell cycle (Georgetown, Tex.) (2015-04-04)
Xiaoyu Xia, Ankur Gholkar, Silvia Senese, Jorge Z Torres
ABSTRACT
Leucine carboxyl methyltransferase-1 (LCMT1) and protein phosphatase methylesterase-1 (PME-1) are essential enzymes that regulate the methylation of the protein phosphatase 2A catalytic subunit (PP2AC). LCMT1 and PME-1 have been linked to the regulation of cell growth and proliferation, but the underlying mechanisms have remained elusive. We show here an important role for an LCMT1-PME-1 methylation equilibrium in controlling mitotic spindle size. Depletion of LCMT1 or overexpression of PME-1 led to long spindles. In contrast, depletion of PME-1, pharmacological inhibition of PME-1 or overexpression of LCMT1 led to short spindles. Furthermore, perturbation of the LCMT1-PME-1 methylation equilibrium led to mitotic arrest, spindle assembly checkpoint activation, defective cell divisions, induction of apoptosis and reduced cell viability. Thus, we propose that the LCMT1-PME-1 methylation equilibrium is critical for regulating mitotic spindle size and thereby proper cell division.
Sigma-Aldrich
Anti-PP2A Antibody, C subunit, demethylated, clone 4b7, ascites fluid, clone 4b7, Upstate®