MilliporeSigma
  • Clathrin Assembly Defines the Onset and Geometry of Cortical Patterning.
Clathrin Assembly Defines the Onset and Geometry of Cortical Patterning.
Developmental cell (2017-11-22)
Yang Yang, Ding Xiong, Anne Pipathsouk, Orion D Weiner, Min Wu
ABSTRACT
Assembly of the endocytic machinery is a constitutively active process that is important for the organization of the plasma membrane, signal transduction, and membrane trafficking. Existing research has focused on the stochastic nature of endocytosis. Here, we report the emergence of the collective dynamics of endocytic proteins as periodic traveling waves on the cell surface. Coordinated clathrin assembly provides the earliest spatial cue for cortical waves and sets the direction of propagation. Surprisingly, the onset of clathrin waves, but not individual endocytic events, requires feedback from downstream factors, including FBP17, Cdc42, and N-WASP. In addition to the localized endocytic assembly at the plasma membrane, intracellular clathrin and phosphatidylinositol-3,4-bisphosphate predict the excitability of the plasma membrane and modulate the geometry of traveling waves. Collectively, our data demonstrate the multiplicity of clathrin functions in cortical pattern formation and provide important insights regarding the nucleation and propagation of single-cell patterns.
Sigma-Aldrich
Anti-Clathrin Light Chain Antibody, serum, Chemicon®