MilliporeSigma
  • Activation of the non-receptor tyrosine kinase cSrc in macrophage-rich atherosclerotic plaques of human carotid arteries.
Activation of the non-receptor tyrosine kinase cSrc in macrophage-rich atherosclerotic plaques of human carotid arteries.
Acta histochemica et cytochemica (2008-01-29)
Sono Toi, Noriyuki Shibata, Tatsuo Sawada, Makio Kobayashi, Shinichiro Uchiyama
ABSTRACT
To determine the involvement of the non-receptor tyrosine kinase cSrc in plaque destabilization in carotid atherosclerosis (CAS), which is responsible for cerebral infarction, we performed quantitative and morphological detection of phosphorylated active cSrc (p-cSrc) and histopathological examination in CAS lesions. We examined carotid endarterectomy specimens obtained from 32 CAS patients. Each specimen was used for immunoblot and immunohistochemical analyses of p-cSrc, histopathological analysis, and image analysis of macrophage content. There was a strong positive correlation between cSrc activation on blots and macrophage content on sections. When we defined the macrophage-rich plaque (MRP) and the macrophage-poor plaque (MPP) as having macrophage content more and less than 5%, respectively, the p-cSrc density and the occurrence of plaque hemorrhage and thrombus formation were significantly increased in the MRP group (n=18) compared to the MPP group (n=14). p-cSrc immunoreactivity was localized in lesional endothelial cells, macrophages, and smooth muscle cells, which contained proinflammatory substances: the upstream oxidized low density lipoprotein, tissue factor and osteopontin, and the downstream active forms of extracellular signal-activated kinase and p38 and nuclear factor-kappaB. Our results suggest that cSrc activation in lesional cells contributes to plaque destabilization in CAS via persistent inflammation.
Sigma-Aldrich
Anti-phospho-Src (Tyr416) Antibody, clone 9A6, clone 9A6, Upstate®, from mouse