MilliporeSigma
  • Effects of the histamine H3-agonist (R)-alpha-methylhistamine and the antagonist thioperamide on histamine metabolism in the mouse and rat brain.
Effects of the histamine H3-agonist (R)-alpha-methylhistamine and the antagonist thioperamide on histamine metabolism in the mouse and rat brain.
Journal of neurochemistry (1989-05-01)
R Oishi, Y Itoh, M Nishibori, K Saeki
ABSTRACT
To study the feedback control by histamine (HA) H3-receptors on the synthesis and release of HA at nerve endings in the brain, the effects of a potent and selective H3-agonist, (R)-alpha-methylhistamine, and an H3-antagonist, thioperamide, on the pargyline-induced accumulation of tele-methylhistamine (t-MH) in the brain of mice and rats were examined in vivo. (R)-alpha-Methylhistamine dihydrochloride (6.3 mg free base/kg, i.p.) and thioperamide (2 mg/kg, i.p.), respectively, significantly decreased and increased the steady-state t-MH level in the mouse brain, whereas these compounds produced no significant changes in the HA level. When administered to mice immediately after pargyline (65 mg/kg, i.p.), (R)-alpha-methylhistamine (3.2 mg/kg, i.p.) inhibited the pargyline-induced increase in the t-MH level almost completely during the first 2 h after treatment. Thioperamide (2 mg/kg, i.p.) enhanced the pargyline-induced t-MH accumulation by approximately 70% 1 and 2 h after treatment. Lower doses of (R)-alpha-methylhistamine (1.3 mg/kg) and thioperamide (1 mg/kg) induced significant changes in the pargyline-induced t-MH accumulation in the mouse brain. In the rat, (R)-alpha-methylhistamine (3.2 mg/kg, i.p.) and thioperamide (2 mg/kg, i.p.) also affected the pargyline-induced t-MH accumulation in eight brain regions and the effects were especially marked in the cerebral cortex and amygdala. These results indicate that these compounds have potent effects on HA turnover in vivo in the brain.
Sigma-Aldrich
Thioperamide maleate salt, solid
Sigma-Aldrich
(R)(−)-α-Methylhistamine dihydrochloride, solid, ≥98% (HPLC)