MilliporeSigma
  • Chemical Modification of Poly(1-Trimethylsylil-1-Propyne) for the Creation of Highly Efficient CO2-Selective Membrane Materials.
Chemical Modification of Poly(1-Trimethylsylil-1-Propyne) for the Creation of Highly Efficient CO2-Selective Membrane Materials.
Materials (Basel, Switzerland) (2019-08-31)
Viktoriya Polevaya, Viktoriya Geiger, Galina Bondarenko, Sergey Shishatskiy, Valeriy Khotimskiy
ABSTRACT
The work is devoted to the chemical modification of a polymer that is promising for the creation of gas separation membranes, aimed at increasing the selectivity with respect to CO2. The introduction of ionic liquids into the structure of poly(1-trimethylsilyl-1-propyne) is realized by a two-step process: bromination of the initial polymer with N-bromosuccinimide and subsequent addition of tertiary amine (N-butylimidazole) to it. Depending on the process conditions, the method allows polymers with different contents of the ionic liquid to be obtained. The obtained polymers show good film-forming properties and thermal stability. Depending on the content of the ionic liquid in the polymer matrix, the resistance to aliphatic alicyclic to the majority of halogenated, as well as aromatic hydrocarbons, increases. With an increase of the ionic liquid content in the polymer, the ideal selectivities of CO2/N2 and CO2/CH4 gas pairs increases while maintaining a high level of permeability.
Sigma-Aldrich
1-Butylimidazole, 98%