MilliporeSigma
  • Identification and characterization of the familial adenomatous polyposis coli gene.
Identification and characterization of the familial adenomatous polyposis coli gene.
Cell (1991-08-09)
J Groden, A Thliveris, W Samowitz, M Carlson, L Gelbert, H Albertsen, G Joslyn, J Stevens, L Spirio, M Robertson
ABSTRACT
DNA from 61 unrelated patients with adenomatous polyposis coli (APC) was examined for mutations in three genes (DP1, SRP19, and DP2.5) located within a 100 kb region deleted in two of the patients. The intron-exon boundary sequences were defined for each of these genes, and single-strand conformation polymorphism analysis of exons from DP2.5 identified four mutations specific to APC patients. Each of two aberrant alleles contained a base substitution changing an amino acid to a stop codon in the predicted peptide; the other mutations were small deletions leading to frameshifts. Analysis of DNA from parents of one of these patients showed that his 2 bp deletion is a new mutation; furthermore, the mutation was transmitted to two of his children. These data have established that DP2.5 is the APC gene.
Sigma-Aldrich
Anti-APC (Ab-7) Mouse mAb (CC-1)