MilliporeSigma
  • Syndecan 1 is a critical mediator of macropinocytosis in pancreatic cancer.
Syndecan 1 is a critical mediator of macropinocytosis in pancreatic cancer.
Nature (2019-03-29)
Wantong Yao, Johnathon L Rose, Wei Wang, Sahil Seth, Hong Jiang, Ayumu Taguchi, Jintan Liu, Liang Yan, Avnish Kapoor, Pingping Hou, Ziheng Chen, Qiuyun Wang, Luigi Nezi, Zhaohui Xu, Jun Yao, Baoli Hu, Piergiorgio F Pettazzoni, I Lin Ho, Ningping Feng, Vandhana Ramamoorthy, Shan Jiang, Pingna Deng, Grace J Ma, Peter Den, Zhi Tan, Shu Xing Zhang, Huamin Wang, Y Alan Wang, Angela K Deem, Jason B Fleming, Alessandro Carugo, Timothy P Heffernan, Anirban Maitra, Andrea Viale, Haoqiang Ying, Samir Hanash, Ronald A DePinho, Giulio F Draetta
ABSTRACT
Pancreatic ductal adenocarcinoma (PDAC) remains recalcitrant to all forms of cancer treatment and carries a five-year survival rate of only 8%1. Inhibition of oncogenic KRAS (hereafter KRAS*), the earliest lesion in disease development that is present in more than 90% of PDACs, and its signalling surrogates has yielded encouraging preclinical results with experimental agents2-4. However, KRAS*-independent disease recurrence following genetic extinction of Kras* in mouse models anticipates the need for co-extinction strategies5,6. Multiple oncogenic processes are initiated at the cell surface, where KRAS* physically and functionally interacts to direct signalling that is essential for malignant transformation and tumour maintenance. Insights into the complexity of the functional cell-surface-protein repertoire (surfaceome) have been technologically limited until recently and-in the case of PDAC-the genetic control of the function and composition of the PDAC surfaceome in the context of KRAS* signalling remains largely unknown. Here we develop an unbiased, functional target-discovery platform to query KRAS*-dependent changes of the PDAC surfaceome, which reveals syndecan 1 (SDC1, also known as CD138) as a protein that is upregulated at the cell surface by KRAS*. Localization of SDC1 at the cell surface-where it regulates macropinocytosis, an essential metabolic pathway that fuels PDAC cell growth-is essential for disease maintenance and progression. Thus, our study forges a mechanistic link between KRAS* signalling and a targetable molecule driving nutrient salvage pathways in PDAC and validates oncogene-driven surfaceome annotation as a strategy to identify cancer-specific vulnerabilities.